missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidophenol, 98%

Product Code. p-4229047 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic Drum
Plastic bottle
Unit Size:
1kg
250g
5g
5kg
This item is not returnable. View return policy

Product Code. 10634942

Brand: Thermo Scientific Acros 102330050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 103-90-2 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-90-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00002328
InChI Key RZVAJINKPMORJF-UHFFFAOYSA-N
Synonym acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol
PubChem CID 1983
ChEBI CHEBI:46195
IUPAC Name N-(4-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC=C(O)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 168.0°C to 172.0°C
Color White
Density 1.2930g/mL
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula CH3CONHC6H4OH
Quantity 5 g
Beilstein 13,46
Merck Index 15,47
Specific Gravity 1.293
Solubility Information Solubility in water: soluble 14g/L in water (20°C). Other solubilities: soluble in ethanol,acetone,alcohol,methanol,,dmf,ethylene dichloride,ethyl acetate,practically insoluble in petr.ether,pentane and,benzene
Formula Weight 151.16
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Acetamidophenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Acetamidophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.