missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidophenol, 98%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar A11240.36


Quantity 500g

 View more versions of this product

Product Code. 11359439

  • £59.10 / 500g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cyclooxygenase inhibitor and widely used analgesic. 4-Acetamidophenol formulated pharmaceutical products are used as antiinfectant, analgesic, anti rheumatic and antipyretic. It is used as an intermediate in organic synthesis, hydrogen peroxide stabilizer and photographic chemicals.

Solubility
Soluble in alcohol, dimethylformamide, ethylene dichloride, acetone and ethyl acetate. Slightly soluble in water and ether.

Notes
Incompatible with strong oxidizing agents, acids and alkali metals.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

103-90-2
151.17
RZVAJINKPMORJF-UHFFFAOYSA-N
1983
N-(4-hydroxyphenyl)acetamide
C8H9NO2
MFCD00002328
acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol
CHEBI:46195
CC(=O)NC1=CC=C(O)C=C1

Specifications

103-90-2
C8H9NO2
500g
14,47
Soluble in alcohol,dimethylformamide,ethylene dichloride,acetone and ethyl acetate. Slightly soluble in water and ether.
CC(=O)NC1=CC=C(O)C=C1
151.17
CHEBI:46195
98%
1.293
MFCD00002328
2208089
acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol
RZVAJINKPMORJF-UHFFFAOYSA-N
N-(4-hydroxyphenyl)acetamide
1983
151.17
4-Acetamidophenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Acetamidophenol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contain: Acetaminophen

SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.