missing translation for 'onlineSavingsMsg'
Learn More

4-(Acetamidomethyl)benzeneboronic acid, 97%

Product Code. 15499357
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499357 1 g 1g
15409367 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499357 Supplier Thermo Scientific Alfa Aesar Supplier No. H52602.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 850568-41-1 | C9H12BNO3 | 193.009 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850568-41-1
Molecular Formula C9H12BNO3
Molecular Weight (g/mol) 193.009
MDL Number MFCD06659818
InChI Key ZMJVNKSOLIUBKO-UHFFFAOYSA-N
Synonym 4-acetamidomethyl phenyl boronic acid, 4-acetamidomethylphenylboronic acid, 4-acetamidomethylphenyl boronic acid, 4-acetamidomethyl benzeneboronic acid, 4-acetylaminomethyl benzeneboronic acid, 4-acetamidomethyl phenylboronic acid, boronic acid,b-4-acetylamino methyl phenyl, 4-acetamidomethylbenzeneboronic acid, pubchem14580
PubChem CID 22309454
IUPAC Name [4-(acetamidomethyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)CNC(=O)C)(O)O

Specifications

Melting Point 278°C to 280°C
Quantity 1 g
Formula Weight 193.01
Percent Purity 97%
Chemical Name or Material 4-(Acetamidomethyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.