missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidobutyric acid, 99%, Thermo Scientific™

Product Code. 11428873
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428873 10 g 10g
11438873 50 g 50g
11448873 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428873 Supplier Thermo Scientific Alfa Aesar Supplier No. A11039.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Acetamidobutyric acid is used as pharmaceutical intermediate.

Solubility
Soluble in water (19.9 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3025-96-5
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.158
MDL Number MFCD00020552
InChI Key UZTFMUBKZQVKLK-UHFFFAOYSA-N
Synonym 4-acetamidobutyric acid, 4-acetylamino butanoic acid, n-acetyl-4-aminobutyric acid, n-acetyl-4-aminobutanoic acid, n-acetyl-gamma-aminobutyrate, butyric acid, 4-acetamido, 4-acetamidobutanoate, 4-acetamidobutyrate, gamma-acetylaminobutyric acid, n4-acetylaminobutanoate
PubChem CID 18189
ChEBI CHEBI:17645
IUPAC Name 4-acetamidobutanoic acid
SMILES CC(=O)NCCCC(=O)O

Specifications

Melting Point 129°C to 131°C
Beilstein 1762288
Solubility Information Soluble in water (19.9mg/ml).
Quantity 10 g
Formula Weight 145.16
Percent Purity 99%
Chemical Name or Material 4-Acetamidobutyric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.