missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidobenzoic acid, 98%

Product Code. 11470633
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470633 250 g 250g
11460633 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470633 Supplier Thermo Scientific Alfa Aesar Supplier No. A12711.30
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 556-08-1 | C9H9NO3 | 179.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-08-1
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00002534
InChI Key QCXJEYYXVJIFCE-UHFFFAOYSA-N
Synonym acedoben, p-acetamidobenzoic acid, 4-acetylamino benzoic acid, 4-carboxyacetanilide, 4-acetylaminobenzoic acid, benzoic acid, 4-acetylamino, p-acetylamino benzoic acid, p-acetoaminobenzoic acid, p-acetaminobenzoic acid, n-acetyl-p-aminobenzoic acid
PubChem CID 19266
ChEBI CHEBI:46171
IUPAC Name 4-acetamidobenzoic acid
SMILES CC(=O)NC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point ∼261°C (decomposition)
Quantity 250 g
Beilstein 390602
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material 4-Acetamidobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.