missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Acetamidobenzenesulfonyl chloride, 98+%

Product Code. p-7081496
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434503 250 g 250g
11424503 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434503 Supplier Alfa Aesar™ Supplier No. A11511.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
N-Acetylsulfanilyl chloride is used in the preparation of sulfanilamide and its derivatives which are intermediates to produce sulfa drugs. They are used in the prevention and treatment of bacterial infections, diabetes mellitus, edema, hypertension, and gout. It is also used as a Pharma raw material. N-Acetylsulfanilyl chloride widely used in the fields of dye, medicine, mainly used for preparation of Sulfanilamide, Sulfanilylureal, Sulfatolamide, Sulphathiourea , Sulfaguanidine and Sulfacetamide and etc.

Solubility
It is slightly in water. But soluble in benzene, chloroform and ether.

Notes
Stable under ordinary conditions. Moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents, Strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-60-8
Molecular Formula C8H8ClNO3S
Molecular Weight (g/mol) 233.67
MDL Number MFCD00007442
InChI Key GRDXCFKBQWDAJH-UHFFFAOYSA-N
Synonym n-acetylsulfanilyl chloride, 4-acetamidobenzene-1-sulfonyl chloride, dagenan chloride, 4-acetylamino benzenesulfonyl chloride, 4-acetamidophenylsulfonyl chloride, benzenesulfonyl chloride, 4-acetylamino, acetylsulfanilyl chloride, p-acetamidobenzenesulfonyl chloride, n-acetylsulphanilyl chloride, p-acetaminobenzenesulfonyl chloride
PubChem CID 8481
SMILES CC(=O)NC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point ∼146°C (decomposition)
Quantity 250 g
UN Number UN3261
Beilstein 746676
Merck Index 14,103
IUPAC Name 4-acetamidobenzene-1-sulfonyl chloride
Formula Weight 233.67
Percent Purity ≥98%
Chemical Name or Material 4-Acetamidobenzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.