missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidobenzenesulfonic Acid, TRC

Product Code. 30138122
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138122 10 g 10g
30005925 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138122 Supplier Toronto Research Chemicals Supplier No. TRCA14187510G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-62-0
Molecular Formula C8 H9 N O4 S
Molecular Weight (g/mol) 215.23
Synonym Benzenesulfonic acid, 4-(acetylamino)-, Sulfanilic acid, N-acetyl- (6CI,7CI,8CI), 4-(Acetylamino)benzenesulfonic acid, 4-Acetamidobenzenesulfonic acid, 4-Acetylaminobenzenesulfonic acid, Acetanilide-p-sulfonic acid, N-Acetylsulfanilic acid, N-p-Acetylanilinosulfonic acid, N4-Acetylsulfanilic acid, p-Acetamidobenzenesulfonic acid, p-Acetylaminobenzenesulfonic acid, 4-(Acetylamino)benzene-1-sulfonic acid
IUPAC Name 4-acetamidobenzenesulfonic acid
SMILES CC(=O)Nc1ccc(cc1)S(=O)(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H9NO4S/c1-6(10)9-7-2-4-8(5-3-7)14(11,12)13/h2-5H,1H3,(H,9,10)(H,11,12,13)
Formula Weight 215.0252
Chemical Name or Material 4-Acetamidobenzenesulfonic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.