missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-TEMPO, free radical, 98+%

Product Code. p-7027021
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455447 1 g 1g
11465447 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455447 Supplier Thermo Scientific Alfa Aesar Supplier No. B23456.03
Add to basket
This item is not returnable. View return policy

CAS: 14691-89-5 | C11H21N2O2 | 213.301 g/mol

4-Acetamido-TEMPO, free radical oxidizes alcohols to carbonyl compounds in the presence of TsOH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Acetamido-TEMPO, free radical oxidizes alcohols to carbonyl compounds in the presence of TsOH.

Solubility
Soluble in ethanol, acetone, acetonitrile, methylene chloride. Insoluble in hexane, ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14691-89-5
Molecular Formula C11H21N2O2
Molecular Weight (g/mol) 213.301
MDL Number MFCD00043593
InChI Key UXBLSWOMIHTQPH-UHFFFAOYSA-N
Synonym 4-acetamido-tempo, 4-acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-acetamido-tempo, free radical, unii-g7abj73l6s, g7abj73l6s, 4-acetylamino-2,2,6,6-tetramethylpiperidin-1-oxyl, pubchem9193, aa-tempo, 4-acetamido-2,2,6,6-tetramethyl-1-piperidinyl oxidanyl
PubChem CID 518988
IUPAC Name N-(1-$l^{1}-oxidanyl-2,2,6,6-tetramethylpiperidin-4-yl)acetamide
SMILES CC(=O)NC1CC(N(C(C1)(C)C)[O])(C)C

Specifications

Melting Point 143°C to 146°C
Quantity 1 g
Beilstein 3546225
Solubility Information Soluble in ethanol,acetone,acetonitrile,methylene chloride. Insoluble in hexane,ether.
Formula Weight 213.3
Percent Purity ≥98%
Chemical Name or Material 4-Acetamido-TEMPO, free radical
compliance-icons
Product Identifier
  • 4-Acetamido-TEMPO, free radical
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4-Acetamido-TEMPO

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.