Learn More
4-Acetamido-TEMPO, free radical, 98+%
Description
4-Acetamido-TEMPO, free radical oxidizes alcohols to carbonyl compounds in the presence of TsOH.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Acetamido-TEMPO, free radical oxidizes alcohols to carbonyl compounds in the presence of TsOH.
Solubility
Soluble in ethanol, acetone, acetonitrile, methylene chloride. Insoluble in hexane, ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, heat.
Specifications
Specifications
| Melting Point | 143°C to 146°C |
| Quantity | 5 g |
| Beilstein | 3546225 |
| Solubility Information | Soluble in ethanol,acetone,acetonitrile,methylene chloride. Insoluble in hexane,ether. |
| Formula Weight | 213.3 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 4-Acetamido-TEMPO, free radical |
Safety and Handling
- 4-Acetamido-TEMPO, free radical
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- MIXTURE LIST-Contain: 4-Acetamido-TEMPO
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.