missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-TEMPO, 99%, Honeywell™ Fluka™

Product Code. 15645830 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15645830 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15645830 Supplier Honeywell-Fluka Supplier No. 557421G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 14691-89-5
Melting Point 143°C to 145°C
Color Orange
Packaging Glass bottle
Molecular Formula C11H21N2O2
MDL Number MFCD00043593
Quantity 1 g
UN Number NONH for all modes of transport
Beilstein 3546225
Synonym 4-acetamido-tempo, 4-acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-acetamido-tempo, free radical, unii-g7abj73l6s, g7abj73l6s, 4-acetylamino-2,2,6,6-tetramethylpiperidin-1-oxyl, pubchem9193, aa-tempo, 4-acetamido-2,2,6,6-tetramethyl-1-piperidinyl oxidanyl
InChI Key UXBLSWOMIHTQPH-UHFFFAOYSA-N
SMILES CC(=O)NC1CC(N(C(C1)(C)C)[O])(C)C
IUPAC Name N-(1-$l^{1}-oxidanyl-2,2,6,6-tetramethylpiperidin-4-yl)acetamide
Molecular Weight (g/mol) 213.301
PubChem CID 518988
Formula Weight 213.3g/mol
Percent Purity 0.99
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamido-TEMPO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.