missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamido-3-nitrobenzoic acid, 97+%, Thermo Scientific™

Product Code. 11434163
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434163 100 g 100g
11424163 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434163 Supplier Thermo Scientific Alfa Aesar Supplier No. A10360.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1539-06-6
Molecular Formula C9H8N2O5
Molecular Weight (g/mol) 224.172
MDL Number MFCD00014703
InChI Key BRQIMWBIZLRLSV-UHFFFAOYSA-N
Synonym 4-acetamido-3-nitrobenzoic acid, 4-acetylamino-3-nitrobenzoic acid, benzoic acid, 4-acetylamino-3-nitro, unii-9w2250fzkc, 4-acetamino-3-nitrobenzoic acid, 4-acetamido-3-nitro-benzoic acid, 4-acetylamino-3-nitro-benzoic acid, pubchem16827, acmc-1bqk5
PubChem CID 73758
SMILES CC(=O)NC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 223°C to 228°C
Quantity 100 g
Beilstein 2217819
Formula Weight 224.17
Percent Purity ≥97%
Chemical Name or Material 4-Acetamido-3-nitrobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.