missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 4μ8C

Description
Selective inhibitor of IRE1α ribonuclease (RNase) activity (IC50 = 60 nM). Blocks substrate access to the active site of IRE1α. Inactivates IRE1α-mediated mRNA degradation. Displays no effect on the IRE1α-related endoribonuclease RNase L.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 204.18 |
| Purity | >98% |
| Molecular Weight (g/mol) | 204.18 |
| Chemical Name or Material | 4μ8C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?