missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 4μ8C

Product Code. 18044992
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18044992 10 mg 10mg
18064362 50 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18044992 Supplier Tocris Bioscience™ Supplier No. 4479/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits IRE1α endoribonuclease activity

Selective inhibitor of IRE1α ribonuclease (RNase) activity (IC50 = 60 nM). Blocks substrate access to the active site of IRE1α. Inactivates IRE1α-mediated mRNA degradation. Displays no effect on the IRE1α-related endoribonuclease RNase L.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14003-96-4
Molecular Formula C11H8O4

Specifications

Quantity 10 mg
Formula Weight 204.18
Purity >98%
Molecular Weight (g/mol) 204.18
Chemical Name or Material 4μ8C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.