missing translation for 'onlineSavingsMsg'
Learn More

4μ8C, MedChemExpress

Product Code. 16453676
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453676 25 mg 25mg
16450318 5 mg 5mg
16417858 10 mg 10mg
16403807 50 mg 50mg
16407676 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16453676 Supplier Medchem Express Supplier No. HY19707
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 4μ8C (IRE1 Inhibitor III) is a small-molecule inhibitor of IRE1α.

4μ8C (IRE1 Inhibitor III) is a small-molecule inhibitor of IRE1α.

Specifications

Chemical Name or Material 4μ8C
CAS 14003-96-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C11H8O4
Quantity 25 mg
Synonym IRE1 Inhibitor III
Solubility Information DMSO : ≥ 27 mg/mL (132.24 mM)
SMILES O=CC1=C(O)C=CC2=C1OC(C=C2C)=O
Molecular Weight (g/mol) 204.18
Formula Weight 204.18
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.