missing translation for 'onlineSavingsMsg'
Learn More

4,8-Dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one (Loratadine Impurity), TRC

Product Code. 30133524
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133524 1 mg 1mg
17931485 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133524 Supplier Toronto Research Chemicals Supplier No. TRCD4341601MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133330-60-6
Molecular Formula C14 H9 Cl2 N O
Molecular Weight (g/mol) 278.13
Synonym 4,8-Dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one, Loratadine Impurity E, Loratadine Impurity 24
IUPAC Name 4,8-dichloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-one
SMILES Clc1ccc2C(=O)c3nccc(Cl)c3CCc2c1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C14H9Cl2NO/c15-9-2-4-10-8(7-9)1-3-11-12(16)5-6-17-13(11)14(10)18/h2,4-7H,1,3H2
Formula Weight 277.0061
Chemical Name or Material 4,8-Dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.