missing translation for 'onlineSavingsMsg'
Learn More

4(7)-Hydroxybenzotriazole, TRC

Product Code. 30406801
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406801 50 mg -
30402710 10 mg 10mg
30401074 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406801 Supplier Toronto Research Chemicals Supplier No. TRCH67255050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26725-51-9
Molecular Formula C6H5N3O
Molecular Weight (g/mol) 135.12
Synonym 4-Hydroxybenzotriazole, 1H-Benzotriazol-7-ol, 1H-Benzotriazol-4-ol (8CI,9CI), Benzotriazol-4-ol (7CI), 1H-1,2,3-Benzotriazol-4-ol, 1H-1,2,3-Benzotriazol-7-ol, 2H-1,2,3-Benzotriazol-4-ol, 4-Hydroxy-1H-benzotriazole, 4-Hydroxybenzotriazole
IUPAC Name 3H-benzotriazol-4-ol
SMILES Oc1cccc2nn[nH]c12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H5N3O/c10-5-3-1-2-4-6(5)8-9-7-4/h1-3,10H,(H,7,8,9)
Formula Weight 135.0433
Chemical Name or Material 4(7)-Hydroxybenzotriazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.