missing translation for 'onlineSavingsMsg'
Learn More

4',7-Dimethoxyisoflavone, MedChemExpress

Product Code. 16491678
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491678 5 mg 5mg
16409268 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491678 Supplier Medchem Express Supplier No. HYN2145
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 4',7-Dimethoxyisoflavone is isolated from the leaves of Albizzia lebbeck, which shows antifungal activity.

4',7-Dimethoxyisoflavone is isolated from the leaves of Albizzia lebbeck, which shows antifungal activity.

Specifications

Chemical Name or Material 4',7-Dimethoxyisoflavone
Physical Form Solid
CAS 1157-39-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C17H14O4
For Use With (Application) COVID-19-immunoregulation
Synonym Dimethoxydaidzein
Solubility Information DMSO : 50 mg/mL (177.12 mM; Need ultrasonic)
SMILES O=C1C(C2=CC=C(OC)C=C2)=COC3=CC(OC)=CC=C13
Molecular Weight (g/mol) 282.29
Formula Weight 282.29
Percent Purity 98.71%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.