missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dichloroquinoline, TRC

Product Code. 30381668
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381668 1 g -
30380942 5 g 5g
30387966 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381668 Supplier Toronto Research Chemicals Supplier No. TRCD4360601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-98-6
Molecular Formula C9 H5 Cl2 N
Molecular Weight (g/mol) 198.05
Synonym 4,7-Dichloroquinoline, Quinoline, 4,7-dichloro-, NSC 593
IUPAC Name 4,7-dichloroquinoline
SMILES Clc1ccc2c(Cl)ccnc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H5Cl2N/c10-6-1-2-7-8(11)3-4-12-9(7)5-6/h1-5H
Formula Weight 196.9799
Chemical Name or Material 4,7-Dichloroquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.