missing translation for 'onlineSavingsMsg'
Learn More

4-(7-Chloro-4-quinolinylamino)-2-(diethylaminomethyl)phenol dihydrochloride dihydrate, 98%, Thermo Scientific Chemicals

Product Code. 15435805
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

An anti-malarial quinoline derivative that acts as an histamine N-methyltransferase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

An anti-malarial quinoline derivative that acts as an histamine N-methyltransferase inhibitor.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6398-98-7
Molecular Formula C20H28Cl3N3O3
Molecular Weight (g/mol) 464.812
MDL Number MFCD00078857
InChI Key YVNAYSHNIILOJS-UHFFFAOYSA-N
Synonym amodiaquin dihydrochloride dihydrate, amodiaquine hcl, amodiaquine hydrochloride, amodiaquine dihydrochloride dihydrate, unii-k6pw2s574l, 4-7-chloroquinolin-4-yl amino-2-diethylamino methyl phenol dihydrochloride dihydrate, amodiaquine hydrochloride usp, cam-aq 1
PubChem CID 64646
ChEBI CHEBI:50652
IUPAC Name 4-[(7-chloroquinolin-4-yl)amino]-2-(diethylaminomethyl)phenol;dihydrate;dihydrochloride
SMILES CCN(CC)CC1=C(C=CC(=C1)NC2=C3C=CC(=CC3=NC=C2)Cl)O.O.O.Cl.Cl

Specifications

Color Yellow
Quantity 25 g
Merck Index 14,572
Solubility Information Soluble in DMSO or methanol
Formula Weight 464.82 (392.33 Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-(7-Chloro-4-quinolinylamino)-2-(diethylaminomethyl)phenol dihydrochloride dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'