missing translation for 'onlineSavingsMsg'
Learn More

4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane, 97%

Product Code. 11475220
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
2 g
Unit Size:
0.1g
0.5g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475220 0.1 g 0.1g
11455220 0.5 g 0.5g
11465220 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475220 Supplier Thermo Scientific Alfa Aesar Supplier No. 044504.77
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31364-42-8 | C16H32N2O5 | 332.44 g/mol

4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane is used as a phase transfer catalysts by transferring ions from one phase to another. It is involved in the synthesis of alkalides and electrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31364-42-8
Molecular Formula C16H32N2O5
Molecular Weight (g/mol) 332.44
MDL Number MFCD00005108
InChI Key HDLXPNDSLDLJHF-UHFFFAOYSA-N
Synonym 4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5 tricosane, cryptating agent 221, kryptofix 221, 4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5. tricosane, cryptofix 221, kryptofix-221, kryptofix r 221, kryptofix™ 221
PubChem CID 123438
IUPAC Name 4,7,13,16,21-pentaoxa-1,10-diazabicyclo[8.8.5]tricosane
SMILES C1COCCN2CCOCCOCCN(CCO1)CCOCC2

Specifications

Color Yellow
Density 1.11 g/mL
Refractive Index 1.501
Quantity 0.1 g
Beilstein 616444
Sensitivity Light sensitive
Solubility Information Miscible with water.
Formula Weight 332.44
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.