missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane
Specifications
Specifications
| Chemical Name or Material | 4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane |
| CAS | 23978-09-8 |
| Analyte or Component Names | 4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H36 N2 O6 |
| Certifications/Compliance | ISO 9001 |
| InChI Formula | InChI=1S/C18H36N2O6/c1-7-21-13-14-24-10-4-20-5-11-25-17-15-22-8-2-19(1)3-9-23-16-18-26-12-6-20/h1-18H2 |
| SMILES | C1COCCN2CCOCCOCCN(CCOCCOCC2)CCO1 |
| IUPAC Name | 4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane |
| Molecular Weight (g/mol) | 376.49 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction