missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(6-Methylsulfonyloxybenzothiophen-2-yl)phenyl Methanesulfonate, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
4-(6-Methylsulfonyloxybenzothiophen-2-yl)phenyl Methanesulfonate
Specifications
Specifications
| Chemical Name or Material | 4-(6-Methylsulfonyloxybenzothiophen-2-yl)phenyl Methanesulfonate |
| CAS | 84449-65-0 |
| Analyte or Component Names | 4-(6-Methylsulfonyloxybenzothiophen-2-yl)phenyl Methanesulfonate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H14 O6 S3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzo[b]thiophene-6-ol, 2-[4-[(methylsulfonyl)oxy]phenyl]-, 6-methanesulfonate, Benzo[b]thiophene-6-ol, 2-[4-[(methylsulfonyl)oxy]phenyl]-, methanesulfonate (9CI), 2-[4-[(Methylsulfonyl)oxy]phenyl]benzo[b]thien-6-yl methanesulfonate |
| InChI Formula | InChI=1S/C16H14O6S3/c1-24(17,18)21-13-6-3-11(4-7-13)15-9-12-5-8-14(10-16(12)23-15)22-25(2,19)20/h3-10H,1-2H3 |
| SMILES | CS(=O)(=O)Oc1ccc(cc1)c2cc3ccc(OS(=O)(=O)C)cc3s2 |
| IUPAC Name | [4-(6-methylsulfonyloxy-1-benzothiophen-2-yl)phenyl] methanesulfonate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction