missing translation for 'onlineSavingsMsg'
Learn More

4-(6-Hydrazino-4-pyrimidinyl)morpholine, Thermo Scientific™

Product Code. 10496794
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10496794 5 g Amber Glass Bottle 5g
10486794 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10496794 Supplier Thermo Scientific Maybridge Supplier No. MO08619DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5767-36-2
Molecular Formula C8H13N5O
Molecular Weight (g/mol) 195.226
InChI Key XWDNLTCJJYYYCI-UHFFFAOYSA-N
Synonym 4-6-hydrazinylpyrimidin-4-yl morpholine, 4-6-hydrazinopyrimidin-4-yl morpholine, morpholine, 4-6-hydrazinyl-4-pyrimidinyl, 4-6-hydrazino-4-pyrimidinyl morpholine, 6-morpholin-4-ylpyrimidine-4-ylhydrazine, 4-6-hydrazino-pyrimidin-4-yl-morpholine, 6-morpholin-4-yl-pyrimidin-4-yl-hydrazine
PubChem CID 13393622
IUPAC Name (6-morpholin-4-ylpyrimidin-4-yl)hydrazine
SMILES C1COCCN1C2=CC(=NC=N2)NN

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 195.22
Percent Purity 95%
Chemical Name or Material 4-(6-Hydrazino-4-pyrimidinyl)morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.