missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diphenyl-2-pyrone, 98%, Thermo Scientific Chemicals

Catalog Number p-7081682
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
17372-52-0
C17H12O2
248.28
MFCD00031018
DDGIHXFDWLCKRZ-UHFFFAOYSA-N
296685
4,6-diphenylpyran-2-one
O=C1OC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
17372-52-0
C17H12O2
248.28
MFCD00031018
DDGIHXFDWLCKRZ-UHFFFAOYSA-N
296685
4,6-diphenylpyran-2-one
O=C1OC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 135°C to 138°C
Odor Odorless
Quantity 1 g
Synonym 4,6-diphenyl-2h-pyran-2-one, 4,6-diphenyl-2-pyrone, 2h-pyran-2-one,4,6-diphenyl, pubchem13218, maybridge1_002884, 4,6-diphenyl-2-pyranone, 2h-pyran-2-one, 4,6-diphenyl
Formula Weight 248.27
Percent Purity 98%
Chemical Name or Material 4,6-Diphenyl-2-pyrone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'