missing translation for 'onlineSavingsMsg'
Learn More

4,6-dimethyl-3-nitropyridin-2(1{H})-one, TRC

Product Code. 30391981
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Product Code. Quantity
30391981 100 mg
30408525 50 mg
30406506 10 mg
3 options
This item is not returnable. View return policy

Product Code. 30391981

Brand: Toronto Research Chemicals TRCH669595100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22934-13-0
Formula Weight 168.0535
InChI Formula InChI=1S/C7H8N2O3/c1-4-3-5(2)8-7(10)6(4)9(11)12/h3H,1-2H3,(H,8,10)
IUPAC Name 4,6-dimethyl-3-nitro-1H-pyridin-2-one
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
SMILES CC1=CC(=C(C(=O)N1)[N+](=O)[O-])C
Synonym 2-Pyridinol, 4,6-dimethyl-3-nitro- (7CI,8CI), 4,6-Dimethyl-3-nitro-2(1H)-pyridinone
Chemical Name or Material 4,6-dimethyl-3-nitropyridin-2(1(H))-one

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.