missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxypyrimidine, 98%

Product Code. 10127163 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10127163 500 g Plastic bottle 500g
10376320 25 g Plastic bottle 25g
10715951 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10127163 Supplier Thermo Scientific Acros Supplier No. 115085000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1193-24-4 | C4H4N2O2 | 112.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-24-4
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00016733
InChI Key DUFGYCAXVIUXIP-UHFFFAOYSA-N
Synonym 4,6-dihydroxypyrimidine, pyrimidine-4,6-diol, 4,6-pyrimidinediol, 6-hydroxy-4 1h-pyrimidinone, 4 1h-pyrimidinone, 6-hydroxy, 4 3h-pyrimidinone, 6-hydroxy, 6-hydroxypyrimidin-4 3h-one, 6-hydroxy-1h-pyrimidin-4-one, 4,6-dihydroxypyrimidin, 6-hydroxy-4-pyrimidinone
PubChem CID 14512
SMILES OC1=CC(=O)NC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 500 g
Beilstein 26,444
Solubility Information Solubility in water: 2.5g/L water (20° c)
IUPAC Name 4-hydroxy-1H-pyrimidin-6-one
Formula Weight 112.09
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 6-Dihydroxypyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 4,6-Dihydroxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.