missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxypyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number 10376320
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1193-24-4
C4H4N2O2
112.09
MFCD00016733
DUFGYCAXVIUXIP-UHFFFAOYSA-N
4,6-dihydroxypyrimidine, pyrimidine-4,6-diol, 4,6-pyrimidinediol, 6-hydroxy-4 1h-pyrimidinone, 4 1h-pyrimidinone, 6-hydroxy, 4 3h-pyrimidinone, 6-hydroxy, 6-hydroxypyrimidin-4 3h-one, 6-hydroxy-1h-pyrimidin-4-one, 4,6-dihydroxypyrimidin, 6-hydroxy-4-pyrimidinone
14512
OC1=CC(=O)NC=N1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1193-24-4
C4H4N2O2
112.09
MFCD00016733
DUFGYCAXVIUXIP-UHFFFAOYSA-N
4,6-dihydroxypyrimidine, pyrimidine-4,6-diol, 4,6-pyrimidinediol, 6-hydroxy-4 1h-pyrimidinone, 4 1h-pyrimidinone, 6-hydroxy, 4 3h-pyrimidinone, 6-hydroxy, 6-hydroxypyrimidin-4 3h-one, 6-hydroxy-1h-pyrimidin-4-one, 4,6-dihydroxypyrimidin, 6-hydroxy-4-pyrimidinone
14512
OC1=CC(=O)NC=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Beilstein 26,444
Solubility Information Solubility in water: 2.5g/L water (20° c)
IUPAC Name 4-hydroxy-1H-pyrimidin-6-one
Formula Weight 112.09
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 6-Dihydroxypyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 4,6-Dihydroxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'