missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, TRC

Product Code. 30137149
Change view
Click to view available options
Quantity:
10 mg
250 mg
5 g
Unit Size:
10mg
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137149 5 g 5g
18040830 250 mg 250mg
18061530 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137149 Supplier Toronto Research Chemicals Supplier No. TRCD4545505G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2465-59-0
Molecular Formula C5 H4 N4 O2
Molecular Weight (g/mol) 152.11
Synonym Oxypurinol, 1H-Pyrazolo[3,4-d]pyrimidine-4,6(5H,7H)-dione, 4H-Pyrazolo[3,4-d]pyrimidine-4,6(5H)-dione, 1,7-dihydro- (6CI), 1H,3H,9H-Alloxanthine, 1H,4H,5H,6H,7H-Pyrazolo[3,4-d]pyrimidine-4,6-dione, 1H-Pyrazolo[3,4-d]pyrimidin-4,6-diol, 4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, 4,6-Dioxopyrazolo[3,4-d]pyrimidine, Alloxanthine, BW 55-5, NSC 76239, Oxipurinol, Oxoallopurinol, Oxyprim
IUPAC Name 1,7-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione
SMILES O=C1NC(=O)c2cn[nH]c2N1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C5H4N4O2/c10-4-2-1-6-9-3(2)7-5(11)8-4/h1H,(H3,6,7,8,9,10,11)
Formula Weight 152.0334
Chemical Name or Material Oxypurinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.