missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, 99%

Product Code. 10584531 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging missing translation for 'unitSize'
10584531 5 g Glass bottle 5g
10123541 1 g Glass bottle 1g
10309003 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10584531 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 339940050
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2465-59-0 | C5H4N4O2 | 152.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2465-59-0
Molecular Formula C5H4N4O2
Molecular Weight (g/mol) 152.11
MDL Number MFCD00056934
InChI Key HXNFUBHNUDHIGC-UHFFFAOYSA-N
Synonym oxypurinol, oxipurinol, alloxanthine, 1h-pyrazolo 3,4-d pyrimidine-4,6-diol, oxoallopurinol, 4,6-dihydroxypyrazolo 3,4-d pyrimidine, 1h-pyrazolo 3,4-d pyrimidine-4,6 5h,7h-dione, oxipurinolum, 1h-pyrazolo 3,4-d pyrimidine-4,6 2h,5h-dione, alloxanthin van
PubChem CID 4644
ChEBI CHEBI:28315
IUPAC Name 1,2-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione
SMILES C1=C2C(=NC(=O)NC2=O)NN1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Formula Weight 152.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4,6-Dihydroxypyrazolo[3,4-d]pyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 4,6-Dihydroxypyrazolo[3,4-d]pyrimidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.