missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, 99%, Thermo Scientific Chemicals

Catalog Number p-3736235
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2465-59-0
C5H4N4O2
152.11
MFCD00056934
HXNFUBHNUDHIGC-UHFFFAOYSA-N
oxypurinol, oxipurinol, alloxanthine, 1h-pyrazolo 3,4-d pyrimidine-4,6-diol, oxoallopurinol, 4,6-dihydroxypyrazolo 3,4-d pyrimidine, 1h-pyrazolo 3,4-d pyrimidine-4,6 5h,7h-dione, oxipurinolum, 1h-pyrazolo 3,4-d pyrimidine-4,6 2h,5h-dione, alloxanthin van
4644
CHEBI:28315
1,2-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione
C1=C2C(=NC(=O)NC2=O)NN1
This item is not returnable. View return policy
2465-59-0
C5H4N4O2
152.11
MFCD00056934
HXNFUBHNUDHIGC-UHFFFAOYSA-N
oxypurinol, oxipurinol, alloxanthine, 1h-pyrazolo 3,4-d pyrimidine-4,6-diol, oxoallopurinol, 4,6-dihydroxypyrazolo 3,4-d pyrimidine, 1h-pyrazolo 3,4-d pyrimidine-4,6 5h,7h-dione, oxipurinolum, 1h-pyrazolo 3,4-d pyrimidine-4,6 2h,5h-dione, alloxanthin van
4644
CHEBI:28315
1,2-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione
C1=C2C(=NC(=O)NC2=O)NN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Formula Weight 152.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4,6-Dihydroxypyrazolo[3,4-d]pyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 4,6-Dihydroxypyrazolo¢3,4-d!pyrimidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'