missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxy-5-nitropyrimidine, 95%, Thermo Scientific™

Product Code. 10251383 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10251383 5 g Glass bottle 5g
10359070 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10251383 Supplier Thermo Scientific Acros Supplier No. 305410050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2164-83-2
Molecular Formula C4H3N3O4
Molecular Weight (g/mol) 157.09
MDL Number MFCD00006110
InChI Key ABTLZAVJDRUDNG-UHFFFAOYSA-N
Synonym 4,6-dihydroxy-5-nitropyrimidine, 5-nitropyrimidine-4,6-diol, 4,6-dihydroxy-5-nitropyrimdine, 5-nitro-4,6-pyrimidinediol, 4 1h-pyrimidinone, 6-hydroxy-5-nitro, 6-hydroxy-5-nitro-3h-pyrimidin-4-one, 6-hydroxy-5-nitro-4 1h-pyrimidinone, 5-nitro-4,6-dihydroxypyrimidine, 4-hydroxy-5-nitro-1h-pyrimidin-6-one, 4 3h-pyrimidinone, 6-hydroxy-5-nitro
PubChem CID 75105
IUPAC Name 6-hydroxy-5-nitro-3,4-dihydropyrimidin-4-one
SMILES OC1=C(C(=O)NC=N1)[N+]([O-])=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point >300°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Formula Weight 157.09
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4, 6-Dihydroxy-5-nitropyrimidine, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.