missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxy-2-methylpyrimidine, 99%

Product Code. p-7080822
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418813 10 g 10g
11428813 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418813 Supplier Thermo Scientific Alfa Aesar Supplier No. A10535.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40497-30-1 | C5H6N2O2 | 126.12 g/mol

4,6-Dihydroxy-2-methylpyrimidine is used in the preparation of 4,6-dichloro-2-methyl-pyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dihydroxy-2-methylpyrimidine is used in the preparation of 4,6-dichloro-2-methyl-pyrimidine.

Solubility
Soluble in sodium hydroxide.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40497-30-1
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006104,MFCD00205627,MFCD20486784
InChI Key BPSGVKFIQZZFNH-UHFFFAOYSA-N
Synonym 4,6-dihydroxy-2-methylpyrimidine, 2-methylpyrimidine-4,6-diol, 2-methyl-4,6-dihydroxypyrimidine, 2-methyl-4,6-pyrimidinediol, 2-methyl-1h,5h-pyrimidine-4,5-dione, 4 1h-pyrimidinone, 6-hydroxy-2-methyl, 2-methyl-4,6-dihydroxy pyrimidine, 6-hydroxy-2-methyl-3h-pyrimidin-4-one, 6-hydroxy-2-methylpyrimidin-4 3h-one, 6-hydroxy-2-methyl-3,4-dihydropyrimidin-4-one
PubChem CID 222672
IUPAC Name 4-hydroxy-2-methyl-1H-pyrimidin-6-one
SMILES CC1=NC(O)=CC(=O)N1

Specifications

Melting Point >360°C
Quantity 10 g
Beilstein 115646
Solubility Information Soluble in sodium hydroxide.
Formula Weight 126.12
Percent Purity 99%
Chemical Name or Material 4,6-Dihydroxy-2-methylpyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.