missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4,6-Dihydroxy-2-methylpyrimidine, 99%

Brand:  Alfa Aesar™ A10535.18

Code : NEW

Additional Details : CAS Number : 40497-30-1 Weight : 0.05000kg

Quantity 50g

 View more versions of this product

Product Code. 11428813

  • £106.70 / 50g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Description

Description

Applications
4,6-Dihydroxy-2-methylpyrimidine is used in the preparation of 4,6-dichloro-2-methyl-pyrimidine.

Solubility
Soluble in sodium hydroxide.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

4,6-Dihydroxy-2-methylpyrimidine
C5H6N2O2
MFCD00006104
2-Methyl-4,6-pyrimidinediol
40497-30-1
126.12
115646

Specifications

4,6-Dihydroxy-2-methylpyrimidine
50g
99%
126.12
115646
>360°C
40497-30-1
C5H6N2O2
MFCD00006104
2-Methyl-4,6-pyrimidinediol
Videos
SDS