missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dihydroxy-2-mercaptopyrimidine, 98%

Product Code. 10755371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10755371 500 g Plastic bottle 500g
10458830 5 g Glass Bottle 5g
10326180 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10755371 Supplier Thermo Scientific Acros Supplier No. 114965000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 504-17-6 | C4H4N2O2S | 144.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-17-6
Molecular Formula C4H4N2O2S
Molecular Weight (g/mol) 144.15
MDL Number MFCD00006674
InChI Key RVBUGGBMJDPOST-UHFFFAOYSA-N
Synonym 2-thiobarbituric acid, thiobarbituric acid, bathyran, austranal, 4,6-dihydroxy-2-thiopyrimidine, 2-thioxodihydropyrimidine-4,6 1h,5h-dione, 2-thio-4,6-dioxypyrimidine, usaf ek-660, 4,6 1h,5h-pyrimidinedione, dihydro-2-thioxo, 2-mercaptobarbituric acid
PubChem CID 2723628
ChEBI CHEBI:33202
IUPAC Name 2-sulfanylidene-1,3-diazinane-4,6-dione
SMILES O=C1CC(=O)NC(=S)N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Loss on Drying 2 % max. (1 g, 105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 500 g
Beilstein 24, 476
Formula Weight 144.15
Percent Purity ≥97.5% (Acid)
Physical Form Liquid
Chemical Name or Material 4, 6-Dihydroxy-2-mercaptopyrimidine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4,6-Dihydroxy-2-mercaptopyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.