missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloropyrimidine, 97%

Product Code. 11397966
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397966 25 g 25g
11387966 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397966 Supplier Thermo Scientific Alfa Aesar Supplier No. L13212.14
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1193-21-1 | C4H2Cl2N2 | 148.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-21-1
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.97
MDL Number MFCD00006109
InChI Key XJPZKYIHCLDXST-UHFFFAOYSA-N
Synonym pyrimidine, 4,6-dichloro, 4,6-dichloro-pyrimidine, 4,6-dichloro pyrimidine, 4,6-dichloro-1,3-diazine, 4,6-dichlorpyrimidin, dichloropyrmidine, dichloropyrimidine, zlchem 225, pubchem6913, 4,6-dichloropyrimdine
PubChem CID 70943
IUPAC Name 4,6-dichloropyrimidine
SMILES ClC1=CC(Cl)=NC=N1

Specifications

Melting Point 64°C to 68°C
Boiling Point 176°C
Quantity 25 g
UN Number UN3261
Beilstein 111195
Formula Weight 148.98
Percent Purity 98%
Chemical Name or Material 4,6-Dichloropyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.