missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloropyrimidine-5-carboxaldehyde, 97%

Product Code. 15470748
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470748 1 g 1g
15480748 5 g 5g
15490748 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15470748 Supplier Thermo Scientific Alfa Aesar Supplier No. H54449.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5305-40-8 | C5H2Cl2N2O | 176.98 g/mol

4,6-Dichloropyrimidine-5-carboxaldehyde is used as a substrate in the synthesis of N-terminal surrogate in amino acid and peptide analogues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dichloropyrimidine-5-carboxaldehyde is used as a substrate in the synthesis of N-terminal surrogate in amino acid and peptide analogues.

Notes
Incompatible with strong oxidizing agents and strong acids. Store in cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5305-40-8
Molecular Formula C5H2Cl2N2O
Molecular Weight (g/mol) 176.98
MDL Number MFCD02257701
InChI Key XQSJHQXYQAUDFC-UHFFFAOYSA-N
Synonym 4,6-dichloro-5-pyrimidinecarbaldehyde, 4,6-dichloro-pyrimidine-5-carbaldehyde, 4,6-dichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 4,6-dichloro, 4,6-dichloro-5-formylpyrimidine, 4,6-dichloro-5-pyrimidinecarboxaldehyde, 4,6-di chloro pyrimidine-5-aldehyde, 4,6-dichlorpyrimidin-5-carbaldehyd, zlchem 524, pubchem2117
PubChem CID 819691
IUPAC Name 4,6-dichloropyrimidine-5-carbaldehyde
SMILES ClC1=NC=NC(Cl)=C1C=O

Specifications

Melting Point 66°C to 71°C
Quantity 1 g
Formula Weight 175.99
Percent Purity 97%
Chemical Name or Material 4,6-Dichloropyrimidine-5-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.