missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-5-fluoropyrimidine, 98%

Product Code. p-7053967
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15478338

Brand: Thermo Scientific Alfa Aesar H61770.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 213265-83-9 | C4HCl2FN2 | 166.96 g/mol

4,6-Dichloro-5-Fluoropyrimidine is an useful intermediate for pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dichloro-5-Fluoropyrimidine is an useful intermediate for pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213265-83-9
Molecular Formula C4HCl2FN2
Molecular Weight (g/mol) 166.96
MDL Number MFCD08056331
InChI Key DGMIGAHDDPJOPN-UHFFFAOYSA-N
Synonym 4,6-dichloro-5-fluoro-pyrimidine, pyrimidine, 4,6-dichloro-5-fluoro, 4,6-dichloro-5-fluoro-1,3-diazine, pubchem18606, acmc-209fj8, ksc494o9j, 4,6-dichloro-5-fluoropyrimidine;, 4,6-dichloro-5-fluoro-pyrimidine;, 5-fluoro-4,6-dichloropyrimidine, 4 3h-pyrimidinone,2-chloro-5-fluoro
PubChem CID 11286675
IUPAC Name 4,6-dichloro-5-fluoropyrimidine
SMILES FC1=C(Cl)N=CN=C1Cl

Specifications

Boiling Point 50°C to 60°C (14 mmHg)
Flash Point 91°C (196°F)
Quantity 1 g
UN Number UN3265
Solubility Information Slightly soluble in water.
Formula Weight 166.97
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4,6-Dichloro-5-fluoropyrimidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.