missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine, 95%, Thermo Scientific™

Product Code. 15486316
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15486316 5 g 5g
15496316 250 mg 250mg
15476316 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15486316 Supplier Thermo Scientific Alfa Aesar Supplier No. H61166.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150728-13-5
Molecular Formula C15H10Cl2N4O2
Molecular Weight (g/mol) 349.171
MDL Number MFCD03839838
InChI Key IZGOBGVYADHVKH-UHFFFAOYSA-N
PubChem CID 10545706
IUPAC Name 4,6-dichloro-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidine
SMILES COC1=CC=CC=C1OC2=C(N=C(N=C2Cl)C3=NC=CC=N3)Cl

Specifications

Quantity 5 g
Synonym 4,6-dichloro-5-2-methoxyphenoxy-2,2'-bipyrimidine, 4,6-dichloro-5-2-methoxyphenoxy-2,2-bipyrimidine, 2,2'-bipyrimidine, 4,6-dichloro-5-2-methoxyphenoxy, 4,6-dichloro-5-2-methoxy-phenoxy-2,2' bipyrimidinyl, 2,2'-bipyrimidine,4,6-dichloro-5-2-methoxyphenoxy, 4,6-dichloro-5-2-methoxyphenoxy-2-pyrimidin-2-yl pyrimidine, 4,6-dichloro-5-2-methoxyphenoxy 2,2'-bipyrimidine, 4,6-dichloro-5-o-methoxyphenoxy-2-2-pyrimidinyl pyrimidine, pubchem17752
Formula Weight 349.17
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.