missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-2-(methylthio)pyrimidine, TRC

Product Code. 30375695
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375695 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375695 Supplier Toronto Research Chemicals Supplier No. TRCD43558050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,6-Dichloro-2-(methylthio)pyrimidine
CAS 6299-25-8
Molecular Formula C5 H4 Cl2 N2 S
Quantity 50 g
Synonym 2-Methylthio-4,6-dichloropyrimidine, 4,6-Dichloro-2-(methylmercapto)pyrimidine, 4,6-Dichloro-2-(methylsulfanyl)pyrimidine, NSC 44560
InChI Formula InChI=1S/C5H4Cl2N2S/c1-10-5-8-3(6)2-4(7)9-5/h2H,1H3
SMILES CSc1nc(Cl)cc(Cl)n1
IUPAC Name 4,6-dichloro-2-methylsulfanylpyrimidine
Molecular Weight (g/mol) 195.07
Formula Weight 193.9472
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.