missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-2-(methylthio)pyrimidine, 98%

Product Code. 10665792 Shop All Thermo Scientific Chemicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
Packaging:
Glass bottle
missing translation for 'unitSize':
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging missing translation for 'unitSize'
10665792 5 g Glass bottle 5g
10791355 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10665792 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 163030050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6299-25-8 | C5H4Cl2N2S | 195.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6299-25-8
Molecular Formula C5H4Cl2N2S
Molecular Weight (g/mol) 195.07
MDL Number MFCD00006086
InChI Key FCMLONIWOAGZJX-UHFFFAOYSA-N
Synonym 4,6-dichloro-2-methylthio pyrimidine, 4,6-dichloro-2-methylmercaptopyrimidine, 4,6-dichloro-2-methylsulfanyl-pyrimidine, 4,6-dichloro-2-methylthiopyrimidine, pyrimidine, 4,6-dichloro-2-methylthio, 4,6-dichloro-2-methylsulfanyl pyrimidine, 4,6-dichloropyrimidine methyl sulphide, 2-methylthio-4,6-dichloropyrimidine, pubchem21477
PubChem CID 80531
IUPAC Name 4,6-dichloro-2-methylsulfanylpyrimidine
SMILES CSC1=NC(=CC(=N1)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 38.0°C to 42.0°C
Color Orange or White to Yellow
Boiling Point 135.0°C to 136.0°C (14.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 23, 371
Formula Weight 195.07
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 4, 6-Dichloro-2-(methylthio)pyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 4,6-Dichloro-2-(methylthio)pyrimidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.