missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-2,5-Diformamidopyrimidine, TRC

Product Code. 30180363
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180363 250 mg 250mg
30179903 50 mg 50mg
30179881 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180363 Supplier Toronto Research Chemicals Supplier No. TRCD398480250MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116477-30-6
Molecular Formula C6 H4 Cl2 N4 O2
Molecular Weight (g/mol) 235.0276
Synonym Formamide, N-[4,6-dichloro-2-(formylamino)-5-pyrimidinyl]-, Formamide, N,N'-(4,6-dichloro-2,5-pyrimidinediyl)bis- (9CI), N-[4,6-Dichloro-2-(formylamino)-5-pyrimidinyl]formamide, 4,6-Dichloro-2,5-diformamidopyridine, 4,6-Dichloro-2,5-diformamidopyrimidine, N,N'-(4,6-Dichloropyrimidine-2,5-diyl)diformamide
IUPAC Name N-(4,6-dichloro-2-formamidopyrimidin-5-yl)formamide
SMILES Clc1nc(NC=O)nc(Cl)c1NC=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C6H4Cl2N4O2/c7-4-3(9-1-13)5(8)12-6(11-4)10-2-14/h1-2H,(H,9,13)(H,10,11,12,14)
Formula Weight 233.9711
Chemical Name or Material 4,6-Dichloro-2,5-diformamidopyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.