missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dibromo-o-xylene, TRC

Product Code. 30380867
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380867 500 mg -
30380226 250 mg -
30387686 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380867 Supplier Toronto Research Chemicals Supplier No. TRCD512095500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51209-82-6
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
Synonym 1,5-Dibromo-2,3-dimethylbenzene, 3,5-Dibromo-o-xylene
SMILES Cc1cc(Br)cc(Br)c1C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H8Br2/c1-5-3-7(9)4-8(10)6(5)2/h3-4H,1-2H3
Formula Weight 261.8993
Chemical Name or Material 4,6-Dibromo-o-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.