missing translation for 'onlineSavingsMsg'
Learn More

4,6-Decadiyne, 97%

Product Code. 15490308
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490308 5 g 5g
15480308 1 g 1g
15400318 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490308 Supplier Thermo Scientific Alfa Aesar Supplier No. H53467.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16387-71-6 | C10H14 | 134.222 g/mol

6-Decen-6-yne is prepared from 4,6-Decadiyne. PDA(poly 4,6-Decadiyne) is used in preparation of thin films. 4,6-decadiyne is used as a substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Decen-6-yne is prepared from 4,6-Decadiyne. PDA(poly 4,6-Decadiyne) is used in preparation of thin films. 4,6-decadiyne is used as a substrate.

Solubility
Immiscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16387-71-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00041645
InChI Key LIWZSNTUMSGWTF-UHFFFAOYSA-N
Synonym 4,6-decadiyne, acmc-1c48i
PubChem CID 140067
IUPAC Name deca-4,6-diyne
SMILES CCCC#CC#CCCC

Specifications

Density 0.82
Boiling Point 96°C (15 mmHg)
Linear Formula CH3(CH2)2C≡CC≡C(CH2)2CH3
Quantity 5 g
Solubility Information Immiscible with water.
Formula Weight 134.22
Percent Purity 97%
Chemical Name or Material 4,6-Decadiyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.