missing translation for 'onlineSavingsMsg'
Learn More

4-(6-Bromo-2-benzothiazolyl)benzenamine, MedChemExpress

Product Code. 16420797
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16420797 50 mg 50mg
16404488 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16420797 Supplier Medchem Express Supplier No. HY111514

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 4-(6-Bromo-2-benzothiazolyl)benzenamine is a β-amyloid PET (positron emission tomography) tracer that can be used in the diagnosis of neurological diseases, such as Alzheimer's and Down's syndrome.

4-(6-Bromo-2-benzothiazolyl)benzenamine is a β-amyloid PET (positron emission tomography) tracer that can be used in the diagnosis of neurological diseases, such as Alzheimer's and Down's syndrome.

Specifications

Chemical Name or Material 4-(6-Bromo-2-benzothiazolyl)benzenamine
Physical Form Powder
CAS 566169-97-9
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C13H9BrN2S
For Use With (Application) Neuroscience-Neurodegeneration
SMILES NC1=CC=C(C2=NC3=CC=C(Br)C=C3S2)C=C1
Molecular Weight (g/mol) 305.19
Formula Weight 305.19
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.