missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Bis(diphenylphosphino)phenoxazine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
4,6-Bis(diphenylphosphino)phenoxazine used as an intermediate in the manufacture of pharmaceuticals, agrochemicals, dyestuffs, fine chemicals, medical and material synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,6-Bis(diphenylphosphino)phenoxazine used as an intermediate in the manufacture of pharmaceuticals, agrochemicals, dyestuffs, fine chemicals, medical and material synthesis.
Solubility
Miscible with water.
Notes
Incompatible with oxidizing agents.
4,6-Bis(diphenylphosphino)phenoxazine used as an intermediate in the manufacture of pharmaceuticals, agrochemicals, dyestuffs, fine chemicals, medical and material synthesis.
Solubility
Miscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
Melting Point | 251°C to 254°C |
Quantity | 250 mg |
Solubility Information | Miscible with water. |
Formula Weight | 551.55 |
Percent Purity | ≥98% |
Chemical Name or Material | 4,6-Bis(diphenylphosphino)phenoxazine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'