missing translation for 'onlineSavingsMsg'
Learn More

4-(6-Amino-3-pyridyl)-1-Boc-piperazine, 97%

Product Code. 15430638
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430638 5 g 5g
15440638 250 mg 250mg
15420638 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15430638 Supplier Thermo Scientific Alfa Aesar Supplier No. H54315.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 571188-59-5 | C14H22N4O2 | 278.36 g/mol

4-(6-Amino-3-pyridyl)-1-Boc-piperazine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(6-Amino-3-pyridyl)-1-Boc-piperazine is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 571188-59-5
Molecular Formula C14H22N4O2
Molecular Weight (g/mol) 278.36
MDL Number MFCD11594962
InChI Key RMULRXHUNOVPEI-UHFFFAOYSA-N
PubChem CID 11737525
IUPAC Name tert-butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C1=CN=C(N)C=C1

Specifications

Quantity 5 g
Beilstein 10173494
Sensitivity Air sensitive
Synonym tert-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, 1-boc-4-6-aminopyridin-3-yl piperazine, tert-butyl 4-6-amino-3-pyridyl piperazine-1-carboxylate, 1-boc-4-6-amino-3-pyridyl piperazine, 4-6-amino-3-pyridyl-1-boc-piperazine, 1-piperazinecarboxylic acid, 4-6-amino-3-pyridinyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-6-amino-3-pyridyl piperazine, t-butyl 4-6-aminopyridin-3-yl piperazine-1-carboxylate, tert-butyl4-6-aminopyridin-3-yl piperazine-1-carboxylate, 4-6-amino-3-pyridyl piperazine-1-carboxylic acid tert-butyl ester
Formula Weight 278.35
Percent Purity 97%
Chemical Name or Material 4-(6-Amino-3-pyridyl)-1-Boc-piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.