missing translation for 'onlineSavingsMsg'
Learn More

4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10685187
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10685187

Brand: Thermo Scientific Maybridge MAY00317DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 874838-96-7
Assay Percent Range 97%
Molecular Formula C12H7ClF3NO3S
MDL Number MFCD07774162
Quantity 1 g
Synonym 4-5-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-5-trifluoromethyl-2-pyridinyl oxy benzenesulfonoyl chloride, 4-5-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, 4-5-trifluoromethyl-2-pyridinyl-oxy benzenesulfonoyl chloride, 4-5-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride, trifluoromethylpyridinyloxybenzenesulfonoylchloride, 4-5-trifluoromethyl-2-pyridyloxy benzenesulfonyl chloride, chloro 4-5-trifluoromethyl 2-pyridyloxy phenyl sulfone
InChI Key XEMAYRPWJCLKBQ-UHFFFAOYSA-N
SMILES FC(F)(F)C1=CN=C(OC2=CC=C(C=C2)S(Cl)(=O)=O)C=C1
IUPAC Name 4-[5-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
Molecular Weight (g/mol) 337.70
PubChem CID 24213828
Formula Weight 337.7
Chemical Name or Material 4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulfonyl chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.