missing translation for 'onlineSavingsMsg'
Learn More

4-{[5-(Trifluoromethyl)pyridin-2-yl]amino}butanoic Acid, TRC

Product Code. 30455944
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455944 25 mg 25mg
30456212 50 mg 50mg
30451668 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455944 Supplier Toronto Research Chemicals Supplier No. TRCT90113525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 923121-46-4
Molecular Formula C10H11N2O2F3
Molecular Weight (g/mol) 248.2
IUPAC Name 4-[[5-(trifluoromethyl)pyridin-2-yl]amino]butanoic acid
SMILES OC(=O)CCCNc1ccc(cn1)C(F)(F)F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H11F3N2O2/c11-10(12,13)7-3-4-8(15-6-7)14-5-1-2-9(16)17/h3-4,6H,1-2,5H2,(H,14,15)(H,16,17)
Formula Weight 248.0773
Chemical Name or Material 4-([5-(Trifluoromethyl)pyridin-2-yl]amino)butanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.