missing translation for 'onlineSavingsMsg'
Learn More

4-[5-(Trifluoromethyl)pyrid-2-yl]benzonitrile, 97%, Thermo Scientific™

Catalog Number 10575364
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
892501-99-4
C13H7F3N2
248.21
MFCD09064969
DFPKSXZRVOQLJC-UHFFFAOYSA-N
4-5-trifluoromethyl pyridin-2-yl benzonitrile, 4-5-trifluoromethyl pyrid-2-yl benzonitrile, 4-5-trifluoromethyl-2-pyridinyl benzonitrile, benzonitrile,4-5-trifluoromethyl-2-pyridinyl, 4-5-trifluoromethyl-2-pyridyl benzenecarbonitrile, 4-5-trifluoromethyl pyridin-2-yl benzenecarbonitrile
24229535
4-[5-(trifluoromethyl)pyridin-2-yl]benzonitrile
FC(F)(F)C1=CN=C(C=C1)C1=CC=C(C=C1)C#N
This item is not returnable. View return policy
892501-99-4
C13H7F3N2
248.21
MFCD09064969
DFPKSXZRVOQLJC-UHFFFAOYSA-N
4-5-trifluoromethyl pyridin-2-yl benzonitrile, 4-5-trifluoromethyl pyrid-2-yl benzonitrile, 4-5-trifluoromethyl-2-pyridinyl benzonitrile, benzonitrile,4-5-trifluoromethyl-2-pyridinyl, 4-5-trifluoromethyl-2-pyridyl benzenecarbonitrile, 4-5-trifluoromethyl pyridin-2-yl benzenecarbonitrile
24229535
4-[5-(trifluoromethyl)pyridin-2-yl]benzonitrile
FC(F)(F)C1=CN=C(C=C1)C1=CC=C(C=C1)C#N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 248.21
Percent Purity 97%
Chemical Name or Material 4-[5-(Trifluoromethyl)pyrid-2-yl]benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'