missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride, 97%

Product Code. 15458385
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458385 250 mg 250mg
15438385 1 g 1g
15448385 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15458385 Supplier Thermo Scientific Alfa Aesar Supplier No. H33283.MD
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 874838-96-7 | C12H7ClF3NO3S | 337.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874838-96-7
Molecular Formula C12H7ClF3NO3S
Molecular Weight (g/mol) 337.70
MDL Number MFCD07774162
InChI Key XEMAYRPWJCLKBQ-UHFFFAOYSA-N
Synonym 4-5-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-5-trifluoromethyl-2-pyridinyl oxy benzenesulfonoyl chloride, 4-5-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, 4-5-trifluoromethyl-2-pyridinyl-oxy benzenesulfonoyl chloride, 4-5-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride, trifluoromethylpyridinyloxybenzenesulfonoylchloride, 4-5-trifluoromethyl-2-pyridyloxy benzenesulfonyl chloride, chloro 4-5-trifluoromethyl 2-pyridyloxy phenyl sulfone
PubChem CID 24213828
IUPAC Name 4-[5-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
SMILES FC(F)(F)C1=CN=C(OC2=CC=C(C=C2)S(Cl)(=O)=O)C=C1

Specifications

Melting Point 51°C to 53°C
Quantity 250 mg
UN Number UN3261
Formula Weight 337.71
Percent Purity 97%
Chemical Name or Material 4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.